Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021518.3 | NP_067493.1 | 8 | 794 | ||
GenBank mRNA | AB232585.1 | - | 8 | 575 | ||
AK042862.1 | - | 8 | 764 | |||
AK079071.1 | - | 8 | 795 | |||
AK135421.1 | - | 8 | 778 | |||
AK146179.1 | - | 8 | 779 | |||
AK150030.1 | - | 8 | 826 | |||
AK159607.1 | - | 8 | 815 | |||
AK160333.1 | - | 8 | 795 | |||
AK163320.1 | - | 8 | 777 | |||
AK184956.1 | - | 4 | 299 | |||
AK208722.1 | - | 4 | 298 | |||
AK212917.1 | - | 4 | 320 | |||
BC058382.1 | - | 8 | 792 | 6820798 | ||
X95403.1 | - | 8 | 782 |