Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290749.1 | NP_001277678.1 | 3 | 654 | ||
NM_001290750.1 | NP_001277679.1 | 3 | 654 | |||
NM_021540.4 | NP_067515.2 | 3 | 654 | |||
GenBank mRNA | AF171875.1 | - | 3 | 622 | ||
AK011088.1 | - | 3 | 639 | |||
AK166468.1 | - | 3 | 604 | |||
AK220529.1 | - | 3 | 624 | |||
BC018199.1 | - | 3 | 648 | 4017435 | ||
BC048901.1 | - | 3 | 637 | 6390079 |