Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1galt1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021550.3 | NP_067525.1 | 2 | 837 | ||
GenBank mRNA | AB030184.1 | - | 2 | 749 | ||
AB091728.1 | - | 1 | 665 | |||
AK005118.1 | - | 2 | 768 | |||
AK008040.1 | - | 2 | 784 | |||
AY159320.1 | - | 2 | 752 | |||
BC029909.1 | - | 2 | 803 | 3154812 |