Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021559.2 | NP_067534.2 | 2 | 165 | ||
GenBank mRNA | AB221566.1 | - | 1 | 24 | ||
AF149093.1 | - | 1 | 24 | |||
AK146726.1 | - | 2 | 163 | |||
AY832930.1 | - | 2 | 204 | |||
BC011345.1 | - | 2 | 208 | 3992743 | ||
BC054832.1 | - | 2 | 165 | 6467818 |