Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021567.5 | NP_067542.3 | 10, 11 | 924 | ||
GenBank mRNA | AK004580.1 | - | 10, 11 | 873 | ||
AK005383.1 | - | 10, 11 | 873 | |||
AK077632.1 | - | 10, 11 | 874 | |||
AK082005.1 | - | 10, 11 | 874 | |||
BC010694.1 | - | 10, 11 | 813 | 2654291 |