Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021608.3 | NP_067621.3 | 1, 2 | 146 | ||
GenBank mRNA | AF190795.1 | - | 1, 2 | 117 | ||
AK010238.1 | - | 1, 2 | 126 | |||
AK051957.1 | - | 1, 2 | 85 | |||
AK076100.1 | - | 1, 2 | 119 | |||
AK076738.1 | - | 1, 2 | 146 | |||
AK146123.1 | - | 1, 2 | 112 | |||
AK151193.1 | - | 1, 2 | 118 | |||
AK151876.1 | - | 1, 2 | 118 | |||
AK168983.1 | - | 1, 2 | 126 | |||
AK187397.1 | - | 1, 2 | 63 | |||
BC010834.1 | - | 1, 2 | 115 | 4235312 |