Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021608.3 | NP_067621.3 | 2, 3 | 205 | ||
GenBank mRNA | AF190795.1 | - | 2, 3 | 176 | ||
AK010238.1 | - | 2, 3 | 185 | |||
AK051957.1 | - | 2, 3 | 144 | |||
AK076100.1 | - | 2, 3 | 178 | |||
AK076738.1 | - | 2, 3 | 205 | |||
AK145958.1 | - | 1, 2 | 1156 | |||
AK146123.1 | - | 2, 3 | 171 | |||
AK151193.1 | - | 2, 3 | 177 | |||
AK151876.1 | - | 2, 3 | 177 | |||
AK168983.1 | - | 2, 3 | 185 | |||
AK187397.1 | - | 2, 3 | 122 | |||
BC005671.1 | - | 1, 2 | 1179 | 3153175 | ||
BC010834.1 | - | 2, 3 | 174 | 4235312 | ||
BC092273.1 | - | 1, 2 | 1158 |