Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024177.3 | NP_077139.2 | 8 | 1084 | ||
GenBank mRNA | AK088343.1 | - | 8 | 1024 | ||
AK151598.1 | - | 8 | 1023 | |||
AK159033.1 | - | 8 | 1085 | |||
BC002319.1 | - | 7 | 1060 | 3593984 | ||
BC030739.1 | - | 8 | 1011 | 1493919 |