Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il21r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021887.2 | NP_068687.1 | 8, 9 | 1295 | ||
GenBank mRNA | AB049137.1 | - | 8, 9 | 1187 | ||
AF254068.1 | - | 8, 9 | 999 | |||
AF269134.1 | - | 7, 8 | 857 | |||
AF279436.1 | - | 8, 9 | 1247 | |||
AF477982.1 | - | 7, 8 | 873 | |||
AF477983.1 | - | 7, 8 | 873 | |||
AF477984.1 | - | 7, 8 | 873 | |||
AF477985.1 | - | 7, 8 | 873 | |||
AF477986.1 | - | 7, 8 | 873 | |||
AK150824.1 | - | 8, 9 | 1256 | |||
AK171826.1 | - | 8, 9 | 1234 | |||
AK172032.1 | - | 1267 | ||||
BC057861.1 | - | 7, 8 | 1147 | 5315350 |