Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fads3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021890.3 | NP_068690.3 | 4 | 730 | ||
GenBank mRNA | AA981959.1 | - | 1 | 19 | ||
AB041560.1 | - | 4 | 672 | |||
AK029318.1 | - | 4 | 589 | |||
AK080414.1 | - | 4 | 582 | |||
AK090042.1 | - | 4 | 712 | |||
AK154534.1 | - | 4 | 607 | |||
AK172509.1 | - | 4 | 709 | |||
BC055950.1 | - | 4 | 411 | |||
BC071266.1 | - | 4 | 613 |