Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npvf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021892.1 | NP_068692.1 | 2 | 411 | ||
GenBank mRNA | AB040289.1 | - | 2 | 411 | ||
AF330058.2 | - | 2 | 411 | |||
AK138200.1 | - | 2 | 459 | |||
BC116905.1 | - | 2 | 459 | 40126224 | ||
BC116907.1 | - | 2 | 459 | 40126226 |