Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021896.5 | NP_068696.2 | 5 | 1054 | ||
GenBank mRNA | AF297082.1 | - | 5 | 1029 | ||
AK004815.1 | - | 5 | 988 | |||
AK028558.1 | - | 5 | 977 | |||
AK031305.1 | - | 5 | 967 | |||
BC057327.1 | - | 5 | 964 | 6825960 | ||
BC094310.1 | - | 5 | 1110 | 6838836 |