Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021896.5 | NP_068696.2 | 5 | 787 | ||
GenBank mRNA | AF297082.1 | - | 5 | 762 | ||
AK004815.1 | - | 5 | 721 | |||
AK028558.1 | - | 5 | 710 | |||
AK031305.1 | - | 5 | 700 | |||
BC057327.1 | - | 5 | 697 | 6825960 | ||
BC094310.1 | - | 5 | 843 | 6838836 |