Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53inp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199105.1 | NP_001186034.1 | 5 | 889 | ||
NM_021897.3 | NP_068697.1 | 4 | 851 | |||
GenBank mRNA | AJ131776.1 | - | 4 | 852 | ||
AK142321.1 | - | 4 | 847 | |||
AY034611.1 | - | 5 | 890 | |||
AY034612.1 | - | 4 | 852 | |||
BC132124.1 | - | 3 | 753 | 40130401 | ||
BC138711.1 | - | 3 | 753 | 8861733 | ||
BC145396.1 | - | 4 | 790 |