Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033795.4 | NP_001028967.2 | 7 | 1239 | ||
GenBank mRNA | AK136928.1 | - | 1 | 928 | ||
AK141066.1 | - | 7 | 1241 | |||
BC138958.1 | - | 1 | 515 | 8861980 | ||
BC138959.1 | - | 1 | 515 | 8861981 |