Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153160.4 | NP_694800.1 | 7 | 632 | ||
GenBank mRNA | AJ272345.1 | - | 7 | 588 | ||
AK012953.1 | - | 7 | 646 | |||
AK152289.1 | - | 7 | 633 | |||
AK161588.1 | - | 7 | 627 | |||
AK168912.1 | - | 633 | ||||
AY253297.1 | - | 7 | 641 | |||
BC018382.1 | - | 6 | 570 | 4215774 | ||
BC049231.1 | - | 7 | 608 | 4161178 |