Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200023.1 | NP_001186952.1 | 4 | 379 | ||
GenBank mRNA | AK162670.1 | - | 4 | 380 | ||
BC037669.1 | - | 2 | 91 |