Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm13889 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145034.1 | NP_001138506.1 | 1 | 14 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF467371.1 | - | 271 | Chr. - - [-] on Build |