Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fscb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163271.1 | NP_001156743.1 | 1 | 2039 | ||
GenBank mRNA | EF133693.1 | - | 1 | 1831 |