Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm6460 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037919.3 | NP_001033008.1 | 2 | 247 | ||
GenBank mRNA | BC110565.1 | - | 2 | 247 | 40050213 |
Gene Symbol: Gm5861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC030042.1 | - | 228 |
Gene Symbol: Gm6455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC048648.1 | - | 2 | 200 |
Gene Symbol: Speer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY151400.1 | - | 244 |