Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm6583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039228.4 | NP_001034317.2 | 1 | 294 | ||
GenBank mRNA | AK030210.1 | - | 1 | 295 | ||
BC150956.1 | - | 1 | 151 | 9087867 |