Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160414.1 | NP_001153886.1 | 3 | 218 | ||
GenBank mRNA | AK136630.1 | - | 3 | 191 | ||
AV381411.2 | - | 3 | 232 |