Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039501.2 | NP_001034590.1 | 2 | 386 | ||
GenBank mRNA | AK131754.1 | - | 2 | 387 | ||
BC132270.1 | - | 2 | 377 | 40130547 | ||
BC132272.1 | - | 2 | 377 | 40130549 | ||
DQ437333.1 | - | 2 | 492 |