Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199113.1 | NP_001186042.1 | 14 | 1544 | ||
NM_001199114.1 | NP_001186043.1 | 13 | 1383 | |||
NM_001199115.1 | NP_001186044.1 | 13 | 1483 | |||
NM_001199116.1 | NP_001186045.1 | 13 | 1443 | |||
NM_022880.3 | NP_075018.1 | 12 | 1484 | |||
GenBank mRNA | AF131212.1 | - | 12 | 1485 | ||
AF305501.1 | - | 13 | 1454 | |||
AI046673.1 | - | 1 | 560 | |||
AK004756.1 | - | 13 | 1429 | |||
AK050089.1 | - | 13 | 1468 | |||
AK146265.1 | - | 13 | 1384 | |||
AK149483.1 | - | 13 | 1399 | |||
AK170463.1 | - | 13 | 1415 | |||
AK207160.1 | - | 1 | 13 | |||
AK218017.1 | - | 2 | 140 | |||
BC004828.1 | - | 14 | 1507 | 2631728 | ||
BC006812.1 | - | 12 | 1415 | 3155188 | ||
EU180577.1 | - | 11 | 1147 |