Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022023.2 | NP_071306.2 | 3 | 251 | ||
GenBank mRNA | AB050013.1 | - | 3 | 193 | ||
AK013947.1 | - | 3 | 208 | |||
AK013970.1 | - | 3 | 204 | |||
AK142014.1 | - | 3 | 223 | |||
BC040233.1 | - | 3 | 185 | 5364203 | ||
BC049134.1 | - | 3 | 189 | 6831501 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK034816.1 | - | 1578 | Chr. - - [-] on Build |