Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Perp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022032.4 | NP_071315.1 | 3 | 605 | ||
GenBank mRNA | AF249870.1 | - | 3 | 606 | ||
AK003734.1 | - | 3 | 546 | |||
AK009184.1 | - | 3 | 543 | |||
AK009190.1 | - | 3 | 544 | |||
BC021772.1 | - | 3 | 530 | 3985702 |