Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001036293.2 | NP_001031370.1 | 3, 4 | 309 | ||
GenBank mRNA | AK002810.1 | - | 3, 4 | 311 | ||
AK004200.1 | - | 3, 4 | 231 | |||
AK205591.1 | - | 3, 4 | 189 | |||
BC019448.1 | - | 3, 4 | 309 | 4212132 |