Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor1aip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160180.1 | NP_001153652.1 | 3 | 881 | ||
NM_001160181.1 | NP_001153653.1 | 3 | 829 | |||
NM_001160182.1 | NP_001153654.1 | 3 | 760 | |||
NM_022329.4 | NP_071724.1 | 2 | 769 | |||
GenBank mRNA | AJ251363.1 | - | 2 | 736 | ||
AK003852.1 | - | 2 | 718 | |||
AK032697.1 | - | 2 | 713 | |||
AK150117.1 | - | 3 | 884 | |||
AK150303.1 | - | 3 | 798 | |||
AK151453.1 | - | 3 | 798 | |||
AK152137.1 | - | 3 | 805 | |||
BC039927.1 | - | 2 | 734 | |||
BC053726.1 | - | 3 | 784 | |||
BC060258.1 | - | 3 | 1110 | |||
BC064017.1 | - | 2 | 734 | 3995710 | ||
BC092059.1 | - | 3 | 787 | 4019181 |