Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022378.3 | NP_071773.2 | 2 | 1058 | ||
GenBank mRNA | BC111908.1 | - | 2 | 769 | 40045942 | |
U90538.1 | - | 2 | 1005 | |||
X92592.1 | - | 2 | 943 |