Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301137.1 | NP_001288066.1 | 2 | 983 | ||
NM_022428.3 | NP_071873.2 | 2 | 983 | |||
GenBank mRNA | AB221658.1 | - | 2 | 236 | ||
AJ271055.1 | - | 2 | 247 | |||
AK035140.1 | - | 2 | 986 | |||
AK053729.1 | - | 2 | 871 | |||
BC130029.1 | - | 2 | 446 | 40140906 |