Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019752.3 | NP_062726.3 | 2 | 835 | ||
GenBank mRNA | AF164513.1 | - | 2 | 714 | ||
AF175324.1 | - | 2 | 731 | |||
AK011536.1 | - | 1641 | ||||
AK146338.1 | - | 2 | 726 | |||
AK159188.1 | - | 2 | 655 | |||
BC033276.1 | - | 2 | 695 | |||
BC049880.1 | - | 2 | 835 | 5321861 |