Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039126.2 | NP_001034215.1 | 5 | 1187 | ||
NM_023046.4 | NP_075533.3 | 5 | 988 | |||
GenBank mRNA | AF155352.1 | - | 5 | 939 | ||
AK006515.1 | - | 5 | 989 | |||
AK122456.1 | - | 5 | 1043 | |||
AK153774.1 | - | 5 | 1209 | |||
BC034869.1 | - | 6 | 982 | |||
KC789642.1 | - | 4 | 717 | |||
KC789643.1 | - | 3 | 331 |