Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm7325 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177468.1 | NP_001170939.1 | 2 | 103 | ||
NM_001177469.1 | NP_001170940.1 | 2 | 106 | |||
NM_001177470.1 | NP_001170941.1 | 2 | 147 | |||
GenBank mRNA | CJ043848.1 | - | 2 | 148 | ||
CJ062905.1 | - | 2 | 137 | |||
DQ190000.1 | - | 2 | 103 | |||
W70601.1 | - | 2 | 103 |