Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a3r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023055.2 | NP_075542.2 | 4 | 850 | ||
NM_023449.3 | NP_075938.2 | 3 | 420 | |||
GenBank mRNA | AB026489.1 | - | 4 | 823 | ||
AK002557.1 | - | 3 | 421 | |||
AK039491.1 | - | 4 | 838 | |||
AK075800.1 | - | 4 | 853 | |||
AK075813.1 | - | 4 | 851 | |||
AK084801.1 | - | 4 | 851 | |||
AK169007.1 | - | 4 | 841 | |||
AK170057.1 | - | 4 | 836 | |||
BC065778.1 | - | 3 | 391 | 3467172 | ||
BC070458.1 | - | 4 | 834 | 30603578 |