Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025313.2 | NP_079589.2 | 1 | 163 | ||
GenBank mRNA | AA409366.1 | - | 1 | 167 | ||
AK002330.1 | - | 1 | 113 | |||
AK005082.1 | - | 1 | 107 | |||
AK011374.1 | - | 1 | 84 | |||
AK018011.1 | - | 1 | 109 | |||
AK076064.1 | - | 1 | 88 | |||
AK151151.1 | - | 1 | 98 | |||
AK155285.1 | - | 1 | 147 | |||
AK165932.1 | - | 1 | 112 | |||
BC008273.1 | - | 1 | 72 | 2811419 | ||
CT010177.1 | - | 1 | 12 |