Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cystm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081365.1 | NP_001074834.1 | 2, 3 | 410 | ||
GenBank mRNA | AK002512.1 | - | 283 | |||
AK003470.1 | - | 111 | ||||
AK028217.1 | - | 281 | ||||
AK167679.1 | - | 315 | ||||
BC028765.1 | - | 337 | 1195860 |