Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctex1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025329.3 | NP_079605.1 | 4 | 471 | ||
GenBank mRNA | AK002589.1 | - | 4 | 430 | ||
AK004191.1 | - | 3 | 329 | |||
AK007178.1 | - | 4 | 459 | |||
AK010027.1 | - | 4 | 429 | |||
AK010464.1 | - | 3 | 357 | |||
BC053332.1 | - | 2 | 1791 | |||
BC059716.1 | - | 4 | 472 | 6774264 |