Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025331.2 | NP_079607.1 | 2 | 462 | ||
GenBank mRNA | AK002765.1 | - | 2 | 223 | ||
AK009529.1 | - | 2 | 185 | |||
BC039062.1 | - | 2 | 462 | 5251683 | ||
BC040065.1 | - | 2 | 140 | 4486523 |