Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snupn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178374.3 | NP_848461.1 | 3, 4 | 349 | ||
GenBank mRNA | AK002717.1 | - | 3, 4 | 365 | ||
AK013151.1 | - | 3, 4 | 359 | |||
AK155315.1 | - | 3, 4 | 366 | |||
BC023317.1 | - | 329 | ||||
BC049638.1 | - | 3, 4 | 349 | 6589249 | ||
BC061258.1 | - | 3, 4 | 354 | 30251661 |