Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035123.3 | NP_001030295.1 | 3, 4 | 685 | ||
GenBank mRNA | AK010304.1 | - | 3, 4 | 686 | ||
AK075597.1 | - | 3, 4 | 688 | |||
AK167753.1 | - | 3, 4 | 667 | |||
AK215299.1 | - | 1, 2 | 172 | |||
BC139198.1 | - | 3, 4 | 717 | 8862220 | ||
BC139199.1 | - | 3, 4 | 717 | 8862221 |