Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ypel3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025347.2 | NP_079623.1 | 2 | 226 | ||
NM_026875.2 | NP_081151.2 | 2 | 326 | |||
GenBank mRNA | AB098742.1 | - | 1 | 31 | ||
AF523353.1 | - | 2 | 216 | |||
AK002925.1 | - | 2 | 211 | |||
AK003371.1 | - | 2 | 227 | |||
AK004431.1 | - | 2 | 299 | |||
BC009171.1 | - | 2 | 200 |