Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamtor4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081108.2 | NP_001074577.1 | 3 | 495 | ||
GenBank mRNA | AK002488.1 | - | 3 | 222 | ||
AK003086.1 | - | 3 | 246 | |||
AK003250.1 | - | 3 | 205 | |||
AK010598.1 | - | 3 | 227 | |||
AK146994.1 | - | 3 | 203 | |||
BC056178.1 | - | 2 | 192 | |||
BC144958.1 | - | 3 | 495 | 9053506 |