Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamtor4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081108.2 | NP_001074577.1 | 2 | 340 | ||
GenBank mRNA | AK002488.1 | - | 2 | 67 | ||
AK003086.1 | - | 2 | 91 | |||
AK003250.1 | - | 2 | 50 | |||
AK010598.1 | - | 2 | 72 | |||
BC056178.1 | - | 1 | 37 | |||
BC144958.1 | - | 2 | 340 | 9053506 |