Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025366.3 | NP_079642.1 | 1, 2 | 179 | ||
GenBank mRNA | AK003263.1 | - | 1, 2 | 138 | ||
AK010311.1 | - | 1, 2 | 122 | |||
AK011261.1 | - | 1, 2 | 137 | |||
BC052720.1 | - | 1, 2 | 158 | 6818266 | ||
BC064310.1 | - | 1, 2 | 158 | |||
BC092216.1 | - | 1, 2 | 146 | 30609530 |