Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bccip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025392.2 | NP_079668.2 | 5 | 570 | ||
GenBank mRNA | AK003666.1 | - | 5 | 567 | ||
AK010684.1 | - | 5 | 564 | |||
BC048465.1 | - | 5 | 562 | 6534043 | ||
CD770748.1 | - | 5 | 574 |