Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163525.2 | NP_001156997.1 | 2, 3 | 190 | ||
NM_025393.4 | NP_079669.2 | 2, 3 | 173 | |||
GenBank mRNA | AA726094.1 | - | 2, 3 | 183 | ||
AK003669.1 | - | 2, 3 | 153 | |||
AK019075.1 | - | 2, 3 | 144 | |||
BC025607.1 | - | 1, 2 | 172 | 5151924 |