Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025399.3 | NP_079675.1 | 5 | 593 | ||
GenBank mRNA | AK003991.1 | - | 5 | 600 | ||
AK013174.1 | - | 5 | 549 | |||
AK179685.1 | - | 1 | 49 | |||
AK205876.1 | - | 1 | 49 | |||
BC025444.1 | - | 5 | 595 | 3986887 |