Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mustn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181390.3 | NP_852055.1 | 2, 3 | 484 | ||
GenBank mRNA | AJ277212.1 | - | 2, 3 | 620 | ||
AK003360.1 | - | 2, 3 | 200 | |||
AK003945.1 | - | 2, 3 | 202 | |||
BC116939.1 | - | 2, 3 | 192 | 40126258 | ||
BC116941.1 | - | 2, 3 | 192 | 40126260 |