Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025403.4 | NP_079679.1 | 1, 2 | 140 | ||
GenBank mRNA | AK004120.1 | - | 1, 2 | 141 | ||
AK012875.1 | - | 1, 2 | 97 | |||
BC028497.1 | - | 1, 2 | 95 | 1398559 |