Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110037F02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081183.1 | NP_001074652.1 | 8 | 1301 | ||
GenBank mRNA | AK034232.1 | - | 8 | 1223 | ||
AK052499.1 | - | 8 | 1239 | |||
AK076731.1 | - | 8 | 1271 | |||
AK122509.1 | - | 8 | 1204 | |||
AK155623.1 | - | 8 | 1302 | |||
AK162531.1 | - | 8 | 1270 | |||
AK170075.1 | - | 8 | 1304 | |||
BC033309.1 | - | 8 | 1271 | 5024976 |